Skip to Content
MilliporeSigma
Maltotriose hydrate 95%

Maltotriose hydrate

Maltotriose hydrate 95%
Synonym(s):
O-α-D-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-O-α-D-glucose
Empirical Formula (Hill Notation):
C18H32O16 · xH2O
CAS Number:
Molecular Weight:
504.44 (anhydrous basis)
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing