Skip to Content
MilliporeSigma
Malonyl coenzyme A lithium salt 97% (CP)

Malonyl coenzyme A lithium salt

Malonyl coenzyme A lithium salt 97% (CP)
Synonym(s):
Malonyl coenzyme A lithium salt
Empirical Formula (Hill Notation):
C24H38N7O19P3S · xLi+
CAS Number:
Molecular Weight:
853.58 (free acid basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing