![Mahanimbine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/647/033/86fcde17-89b6-40cb-b7c5-d356cc72ea63/640/86fcde17-89b6-40cb-b7c5-d356cc72ea63.png)
Mahanimbine
![Mahanimbine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/647/033/86fcde17-89b6-40cb-b7c5-d356cc72ea63/640/86fcde17-89b6-40cb-b7c5-d356cc72ea63.png)
Synonym(s):
(+)-3,11-Dihydro-3,5-dimethyl-3-(4-methyl-3-penten-1-yl)pyrano[3,2-a]carbazole, Mahanimbin
Empirical Formula (Hill Notation):
C23H25NO
CAS Number:
Molecular Weight:
331.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing