Skip to Content
MilliporeSigma
Magnesium trifluoromethanesulfonate 97%

Magnesium trifluoromethanesulfonate

Magnesium trifluoromethanesulfonate 97%
Synonym(s):
Magnesium triflate, Trifluoromethanesulfonic acid magnesium salt
Linear Formula:
(CF3SO3)2Mg
CAS Number:
Molecular Weight:
322.44

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing