Skip to Content
MilliporeSigma
Magnesium monoperoxyphthalate hexahydrate technical, ~80% (RT)

Magnesium monoperoxyphthalate hexahydrate

Magnesium monoperoxyphthalate hexahydrate technical, ~80% (RT)
Synonym(s):
Magnesium bis(monoperoxyphthalate) hexahydrate, MMPP
Linear Formula:
(HO2CC6H4CO3)2Mg·6H2O
Molecular Weight:
494.64
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing