Skip to Content
MilliporeSigma
L-Tyrosine hydrochloride ≥98% (TLC)

L-Tyrosine hydrochloride

L-Tyrosine hydrochloride ≥98% (TLC)
Synonym(s):
3-(4-Hydroxyphenyl)-L-alanine, 3-(4-Hydroxyphenyl)-L-alanine hydrochloride
Linear Formula:
HOC6H4CH2CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
217.65

Products

Product Category
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing