Skip to Content
MilliporeSigma
L-Proline benzyl ester hydrochloride 98%

L-Proline benzyl ester hydrochloride

L-Proline benzyl ester hydrochloride 98%
Empirical Formula (Hill Notation):
C12H15NO2 · HCl
CAS Number:
Molecular Weight:
241.71
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing