Skip to Content
MilliporeSigma
L-Phenylalanine-15N endotoxin tested, 98 atom % 15N

L-Phenylalanine-15N

L-Phenylalanine-15N endotoxin tested, 98 atom % 15N
Linear Formula:
C6H5CH2CH(15NH2)CO2H
CAS Number:
Molecular Weight:
166.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing