Skip to Content
MilliporeSigma
Lorazepam Related Compound D certified reference material, pharmaceutical secondary standard

Lorazepam Related Compound D

Lorazepam Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxylic acid
Empirical Formula (Hill Notation):
C15H8Cl2N2O2
CAS Number:
Molecular Weight:
319.14

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade