Skip to Content
MilliporeSigma
Lobeline hydrochloride European Pharmacopoeia (EP) Reference Standard

(−)-Lobeline hydrochloride

Lobeline hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
α-Lobeline hydrochloride, L-Lobeline hydrochloride, (−)-Lobeline hydrochloride
Empirical Formula (Hill Notation):
C22H27NO2 · HCl
CAS Number:
Molecular Weight:
373.92

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Format
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2