Skip to Content
MilliporeSigma
L-Lysine monohydrate BioReagent, suitable for cell culture, from non-animal source

L-Lysine monohydrate

L-Lysine monohydrate BioReagent, suitable for cell culture, from non-animal source
Empirical Formula (Hill Notation):
C6H14N2O2 · H2O
CAS Number:
Molecular Weight:
164.20
EC Number:

Products

Product Category
Biological Source
Color
Formula Weight
Physical Form
Product Line
Markush Class
Markush Group
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1