![L-Lysine acetate salt ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/365/096/44dbc337-66c4-4832-baf6-d8645a660e77/640/44dbc337-66c4-4832-baf6-d8645a660e77.png)
L-Lysine acetate salt
![L-Lysine acetate salt ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/365/096/44dbc337-66c4-4832-baf6-d8645a660e77/640/44dbc337-66c4-4832-baf6-d8645a660e77.png)
Synonym(s):
(S)-2,6-Diaminohexanoic acid
Empirical Formula (Hill Notation):
C6H14N2O2 · C2H4O2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing