Skip to Content
MilliporeSigma
L-Lysine-13C6 hydrochloride 99 atom % 13C, 95% (CP)

L-Lysine-13C6 hydrochloride

L-Lysine-13C6 hydrochloride 99 atom % 13C, 95% (CP)
Synonym(s):
(S)-2,6-Diaminocaproic acid-13C6, 13C Labeled Lys, 13C Labeled lysine
Linear Formula:
H2N(13CH2)413CH(NH2)13CO2H · HCl
CAS Number:
Molecular Weight:
188.60

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing