Skip to Content
MilliporeSigma
L-Leucine-13C6,15N 98 atom % 15N, 98 atom % 13C, 99% chiral purity basis

L-Leucine-13C6,15N

L-Leucine-13C6,15N 98 atom % 15N, 98 atom % 13C, 99% chiral purity basis
Synonym(s):
L-Leucine-ul-13C,15N
Linear Formula:
(13CH3)213CH13CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
138.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique