Skip to Content
MilliporeSigma
Lithium tri-tert-butoxyaluminum hydride 97%

Lithium tri-tert-butoxyaluminum hydride

Lithium tri-tert-butoxyaluminum hydride 97%
Synonym(s):
Lithium aluminum-tri-tert-butoxyhydride
Linear Formula:
LiAlH[OC(CH3)3]3
CAS Number:
Molecular Weight:
254.27
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing