![Lithium trifluoromethanesulfonate 96%](/deepweb/assets/sigmaaldrich/product/structures/174/019/fa4b1ceb-9805-4df5-8aba-fa083d3158b7/640/fa4b1ceb-9805-4df5-8aba-fa083d3158b7.png)
Lithium trifluoromethanesulfonate
![Lithium trifluoromethanesulfonate 96%](/deepweb/assets/sigmaaldrich/product/structures/174/019/fa4b1ceb-9805-4df5-8aba-fa083d3158b7/640/fa4b1ceb-9805-4df5-8aba-fa083d3158b7.png)
Synonym(s):
LiTf, Lithium triflate, Trifluoromethanesulfonic acid lithium salt
Linear Formula:
CF3SO3Li
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing