Skip to Content
MilliporeSigma
Lithium isopropoxide 95%

Lithium isopropoxide

Lithium isopropoxide 95%
Synonym(s):
Lithium 2-propanolate
Linear Formula:
(CH3)2CHOLi
CAS Number:
Molecular Weight:
66.03

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing