
Lithium bis(oxalato)borate

Synonym(s):
LiBOB, Lithium bis(ethanedioato)borate, Lithium bis(oxalate)borate
Linear Formula:
LiB(C2O4)2
CAS Number:
Molecular Weight:
193.79
Products
Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Application
Showing 1-1 of 1