Skip to Content
MilliporeSigma
Lithium acetylacetonate 97%

Lithium acetylacetonate

Lithium acetylacetonate 97%
Synonym(s):
2,4-Pentanedione lithium derivative, Acetylacetone lithium derivative, Li(acac)
Linear Formula:
CH3COCH=C(OLi)CH3
CAS Number:
Molecular Weight:
106.05
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing