Skip to Content
MilliporeSigma
(−)-Limonene oxide, mixture of cis and trans 99%

(−)-Limonene oxide, mixture of cis and trans

(−)-Limonene oxide, mixture of cis and trans 99%
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing