Skip to Content
MilliporeSigma
Lidocaine N-ethyl bromide analytical standard, for drug analysis

Lidocaine N-ethyl bromide

Lidocaine N-ethyl bromide analytical standard, for drug analysis
Synonym(s):
N-(2,6-Dimethylphenylcarbamoylmethyl)triethylammonium bromide, QX-314
Empirical Formula (Hill Notation):
C16H27BrN2O
CAS Number:
Molecular Weight:
343.30

Products

Formula Weight
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing