Skip to Content
MilliporeSigma
L-(−)-Glyceric acid hemicalcium salt monohydrate ≥97.0% (KT)

L-(−)-Glyceric acid hemicalcium salt monohydrate

L-(−)-Glyceric acid hemicalcium salt monohydrate ≥97.0% (KT)
Synonym(s):
(S)-2,3-Dihydroxypropanoic acid hemicalcium salt monohydrate, L-Glyceric acid calcium salt dihydrate
Empirical Formula (Hill Notation):
C6H10O8 · Ca · 2H2O
CAS Number:
Molecular Weight:
286.25

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing