Skip to Content
MilliporeSigma
L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt ≥99.0% (TLC)

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt

L-Glutamic acid γ-(3-carboxy-4-nitroanilide) ammonium salt ≥99.0% (TLC)
Synonym(s):
L-Glutamic acid 5-(3-carboxy-4-nitroanilide) ammonium salt
Linear Formula:
C12H12N3O7NH4
CAS Number:
Molecular Weight:
328.28
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing