![Levoglucosenone ≥95%](/deepweb/assets/sigmaaldrich/product/structures/225/561/db1e7a45-52f4-4a2b-a632-ac6840f7cea7/640/db1e7a45-52f4-4a2b-a632-ac6840f7cea7.png)
Levoglucosenone
![Levoglucosenone ≥95%](/deepweb/assets/sigmaaldrich/product/structures/225/561/db1e7a45-52f4-4a2b-a632-ac6840f7cea7/640/db1e7a45-52f4-4a2b-a632-ac6840f7cea7.png)
Synonym(s):
(1S,5R)-6,8-dioxabicyclo[3.2.1]oct-2-en-4-one, 1,6-anhydro-3,4-dideoxyhex-3-enopyran-2-ulose, 6,8-Dioxabicyclo[3.2.1]oct-2-en-4-one
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing