![Levetiracetam Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/737/315/b0fd4eaa-7184-4f59-a671-1825041e26ad/640/b0fd4eaa-7184-4f59-a671-1825041e26ad.png)
Levetiracetam Related Compound B
![Levetiracetam Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/737/315/b0fd4eaa-7184-4f59-a671-1825041e26ad/640/b0fd4eaa-7184-4f59-a671-1825041e26ad.png)
Synonym(s):
(S)-(+)-2-Aminobutanamide hydrochloride, (S)-2-Aminobutyramide hydrochloride
Linear Formula:
CH3CH2CH(NH2)CONH2 · HCl
CAS Number:
Molecular Weight:
138.60
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2