Skip to Content
MilliporeSigma
Leuprolide Acetate pharmaceutical secondary standard, certified reference material

Leuprolide acetate

Leuprolide Acetate pharmaceutical secondary standard, certified reference material
Synonym(s):
Leuprolide acetate salt, des-Gly10-[D-Leu6]-LH-RH ethylamide, Leuprorelin
Empirical Formula (Hill Notation):
C59H84N16O12 · xC2H4O2
CAS Number:
Molecular Weight:
1209.40 (free base basis)

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade