Skip to Content
MilliporeSigma
Leupeptin hydrochloride microbial, ≥70% (HPLC)

Leupeptin hydrochloride

Leupeptin hydrochloride microbial, ≥70% (HPLC)
Synonym(s):
Acetyl-Leu-Leu-Arg-al hydrochloride
Empirical Formula (Hill Notation):
C20H38N6O4 · HCl
CAS Number:
Molecular Weight:
463.01

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing