Skip to Content
MilliporeSigma
L-Cystine dimethyl ester dihydrochloride ≥95%

L-Cystine dimethyl ester dihydrochloride

L-Cystine dimethyl ester dihydrochloride ≥95%
Linear Formula:
CH3OCOCH(NH2)CH2SSCH2CH(NH2)COOCH3 · 2HCl
CAS Number:
Molecular Weight:
341.28
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing