Skip to Content
MilliporeSigma
L-Cysteine methyl ester hydrochloride ≥95%

L-Cysteine methyl ester hydrochloride

L-Cysteine methyl ester hydrochloride ≥95%
Linear Formula:
HSCH2CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
171.65
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-2 of 2