Skip to Content
MilliporeSigma
(-)-Cinchonidine phyproof® Reference Substance

(-)-Cinchonidine

(-)-Cinchonidine phyproof® Reference Substance
Synonym(s):
Cinchonidine, (−)-Cinchonidine
Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing