![Latanoprost for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/202/034/245140f6-aa52-487e-996a-7ee93ba52afa/640/245140f6-aa52-487e-996a-7ee93ba52afa.png)
Latanoprost for system suitability
![Latanoprost for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/202/034/245140f6-aa52-487e-996a-7ee93ba52afa/640/245140f6-aa52-487e-996a-7ee93ba52afa.png)
Synonym(s):
Latanoprost, Isopropyl (5Z,9α,11α,15R)-9,11,15-trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oate, Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate, Xalatan
Empirical Formula (Hill Notation):
C26H40O5
CAS Number:
Molecular Weight:
432.59
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing