Skip to Content
MilliporeSigma
L-α-Phosphatidylinositol from Glycine max (soybean) ~50% (TLC)

L-α-Phosphatidylinositol from Glycine max (soybean)

L-α-Phosphatidylinositol from Glycine max (soybean) ~50% (TLC)
Synonym(s):
1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol), Afamelanotide
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing