Skip to Content
MilliporeSigma
Lanthanum strontium cobalt ferrite powder, 0.6 μm particle size

Lanthanum strontium cobalt ferrite

Lanthanum strontium cobalt ferrite powder, 0.6 μm particle size
Synonym(s):
LCSF, Lanthanum strontium cobalt iron oxide

Products

Product Category
Sustainability
Feature
Physical Form
Chemical Composition
Application
Particle Size (µm)
Showing 1-2 of 2