Skip to Content
MilliporeSigma
Lanthanum(III) isopropoxide 98%

Lanthanum(III) isopropoxide

Lanthanum(III) isopropoxide 98%
Synonym(s):
Lanthanum isopropylate, Lanthanum(III) tri-isopropoxide, Tris(isopropoxy)lanthanum(III)
Empirical Formula (Hill Notation):
C9H21LaO3
CAS Number:
Molecular Weight:
316.17

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing