Skip to Content
MilliporeSigma
L-Alanine-13C3,15N 98 atom % 15N, 98 atom % 13C

L-Alanine-13C3,15N

L-Alanine-13C3,15N 98 atom % 15N, 98 atom % 13C
Linear Formula:
13CH313CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
93.06

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing