
Lactofen

Synonym(s):
2-Ethoxy-1-methyl-2-oxoethyl 5-(2-chloro-4-trifluoromethylphenoxy)-2-nitrobenzoate, Ethyl O-[5-(2-chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoyl]-DL-lactate
Empirical Formula (Hill Notation):
C19H15ClF3NO7
CAS Number:
Molecular Weight:
461.77
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing