Skip to Content
MilliporeSigma
KU-55933 ≥98% (HPLC)

KU-55933

KU-55933 ≥98% (HPLC)
Synonym(s):
2-(4-Morpholinyl)-6-(1-thianthrenyl)-4H-Pyran-4-one, 2-(Morpholin-4-yl)-6-(thianthren-1-yl)-4H-pyran-4-one
Empirical Formula (Hill Notation):
C21H17NO3S2
CAS Number:
Molecular Weight:
395.49

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing