Skip to Content
MilliporeSigma
KT172 ≥98% (HPLC)

KT172

KT172 ≥98% (HPLC)
Synonym(s):
(2-Benzylpiperidin-1-yl)(4-(2′-methoxy-[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl)methanone, Cravatt Reagent, [4-(2′-Methoxy[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl][2-(phenylmethyl)-1-piperidinyl]-methanone
Empirical Formula (Hill Notation):
C28H28N4O2
CAS Number:
Molecular Weight:
452.55

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing