Skip to Content
MilliporeSigma
KN-62 ≥95%, powder

KN-62

KN-62 ≥95%, powder
Synonym(s):
(S)-5-Isoquinolinesulfonic acid 4-[2-[(5-isoquinolinylsulfonyl)methylamino]-3-oxo-3-(4-phenyl-1-piperazinyl)propyl]phenyl ester, 1-[N,O-bis(5-Isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine
Empirical Formula (Hill Notation):
C38H35N5O6S2
CAS Number:
Molecular Weight:
721.84

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing