Skip to Content
MilliporeSigma
Khellin phyproof® Reference Substance

Khellin

Khellin phyproof® Reference Substance
Synonym(s):
4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one
Empirical Formula (Hill Notation):
C14H12O5
CAS Number:
Molecular Weight:
260.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing