Skip to Content
MilliporeSigma
Kanamycin sulfate from Streptomyces kanamyceticus meets USP testing specifications, powder

Kanamycin sulfate from Streptomyces kanamyceticus

Kanamycin sulfate from Streptomyces kanamyceticus meets USP testing specifications, powder
Synonym(s):
Kanamycin A, Kanamycin sulfate salt
Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Technique