Skip to Content
MilliporeSigma
Kaempferol 3-β-D-glucopyranoside ≥97.0% (HPLC)

Kaempferol 3-β-D-glucopyranoside

Kaempferol 3-β-D-glucopyranoside ≥97.0% (HPLC)
Synonym(s):
3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-glucoside
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38

Products

Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing