K-411 Glass microspheres
Synonym(s):
Glass spheres
Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing