Skip to Content
MilliporeSigma
Isoxadifen-ethyl PESTANAL®, analytical standard

Isoxadifen-ethyl

Isoxadifen-ethyl PESTANAL®, analytical standard
Synonym(s):
Ethyl 4,5-dihydro-5,5-diphenylisoxazol-3-carboxylate
Empirical Formula (Hill Notation):
C18H17NO3
CAS Number:
Molecular Weight:
295.33
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique