![Isopteropodine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/355/967/77bc5f62-eb9a-44b7-ac9f-c9e6acc9e1b8/640/77bc5f62-eb9a-44b7-ac9f-c9e6acc9e1b8.png)
Isopteropodine
![Isopteropodine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/355/967/77bc5f62-eb9a-44b7-ac9f-c9e6acc9e1b8/640/77bc5f62-eb9a-44b7-ac9f-c9e6acc9e1b8.png)
Synonym(s):
7-Isopteropodine
Empirical Formula (Hill Notation):
C21H24N2O4
CAS Number:
Molecular Weight:
368.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing