Skip to Content
MilliporeSigma
Isopropyl myristate Pharmaceutical Secondary Standard; Certified Reference Material

Isopropyl myristate

Isopropyl myristate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Isopropyl tetradecanoate, Myristic acid isopropyl ester, Isopropyl myristate
Linear Formula:
CH3(CH2)12COOCH(CH3)2
CAS Number:
Molecular Weight:
270.45
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique