Skip to Content
MilliporeSigma
Isobutyltriethoxysilane ≥95%

Isobutyltriethoxysilane

Isobutyltriethoxysilane ≥95%
Synonym(s):
Triethoxy(isobutyl)silane, Isobutyltriethoxysilane
Linear Formula:
(CH3)2CHCH2Si(OC2H5)3
CAS Number:
Molecular Weight:
220.38
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing