Skip to Content
MilliporeSigma
Iron(II) ethylenediammonium sulfate tetrahydrate 98%

Iron(II) ethylenediammonium sulfate tetrahydrate

Iron(II) ethylenediammonium sulfate tetrahydrate 98%
Synonym(s):
Ferrous ethylenediammonium sulfate tetrahydrate
Linear Formula:
FeSO4 · NH3CH2CH2NH3SO4 · 4H2O
CAS Number:
Molecular Weight:
382.15
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing