Skip to Content
MilliporeSigma
IR-797 chloride Dye content 70 %

IR-797 chloride

IR-797 chloride Dye content 70 %
Synonym(s):
2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-ethylidene]-1-cyclopenten-1-yl]-ethenyl]-1,3,3-trimethyl-3H-indolium chloride
Empirical Formula (Hill Notation):
C31H34Cl2N2
CAS Number:
Molecular Weight:
505.52

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing