Skip to Content
MilliporeSigma
vibrant-m

Ioxaglic acid

vibrant-m
Synonym(s):
N-(2-Hydroxyethyl)-2,4,6-triiodo-5-{2-[2,4,6-triiodo-3-(N-methylacetamido)-5-(methylcarbamoyl)benzamido]acetamido}isophthalamic acid
Empirical Formula (Hill Notation):
C24H21I6N5O8
CAS Number:
Molecular Weight:
1268.88

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade